siRNA Details
| Gene Symbol: LOC101060091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110174.1 | 5 | 519 | |||
| GenBank mRNA | DB153264.1 | - | 6 | 521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101060091| Gene Symbol: LOC101060091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110174.1 | 5 | 519 | |||
| GenBank mRNA | DB153264.1 | - | 6 | 521 | ||