siRNA Details
| Gene Symbol: C9orf135-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038833.1 | 2 | 328 | |||
| GenBank mRNA | BC039385.1 | - | 2 | 350 | ||
| BG717523.1 | - | 2 | 306 | |||
| BI830190.1 | - | 2 | 328 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA431615.1 | - | 1 | 53 | Chr. 9 - 69819603 - 69820082 [-] on Build GRCh38 | |

C9orf135-AS1
Genomic Map