siRNA Details
| Gene Symbol: LINC01558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026773.1 | 2 | 377 | |||
| GenBank mRNA | AB016901.1 | - | 2 | 389 | ||
| BC027853.1 | - | 2 | 400 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI771211.1 | - | 2 | 399 | Chr. 6 - 167785386 - 167796859 [-] on Build GRCh38 | |

LINC01558
Genomic Map