siRNA Details
| Gene Symbol: LOC102723701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125821.1 | 2 | 781 | |||
| GenBank mRNA | AK091697.1 | - | 2 | 781 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723701| Gene Symbol: LOC102723701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125821.1 | 2 | 781 | |||
| GenBank mRNA | AK091697.1 | - | 2 | 781 | ||