siRNA Details
Gene Symbol: LOC101927727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135633.1 | 1, 2 | 309 | |||
GenBank mRNA | BG617385.1 | - | 1, 2 | 65 | ||
BQ934941.1 | - | 1, 2 | 309 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135633.1 | 1, 2 | 309 | |||
GenBank mRNA | BG617385.1 | - | 1, 2 | 65 | ||
BQ934941.1 | - | 1, 2 | 309 |