siRNA Details
| Gene Symbol: OVCH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073172.1 | 1, 2 | 560 | |||
| GenBank mRNA | BQ058205.1 | - | 1, 2 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OVCH1-AS1| Gene Symbol: OVCH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073172.1 | 1, 2 | 560 | |||
| GenBank mRNA | BQ058205.1 | - | 1, 2 | 560 | ||