siRNA Details
| Gene Symbol: Rap1gapos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007899.1 | - | 2 | 435 | ||
| BQ287036.1 | - | 1 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rap1gapos| Gene Symbol: Rap1gapos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007899.1 | - | 2 | 435 | ||
| BQ287036.1 | - | 1 | 283 | |||