siRNA Details
| Gene Symbol: LOC146880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026899.1 | 12 | 2472 | |||
| GenBank mRNA | AK055342.1 | - | 11 | 2343 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000578492.5 | - | 11 | 2343 | Chr. 17 - 64749663 - 64781626 [-] on Build GRCh38 | |

LOC146880
Genomic Map