siRNA Details
Gene Symbol: LINC01587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126517.1 | 2, 3 | 475 | |||
NR_126518.1 | 2, 3 | 454 | ||||
NR_126519.1 | 3, 4 | 363 | ||||
GenBank mRNA | BC117444.1 | - | 2, 3 | 408 | 40125995 | |
BC117446.1 | - | 2, 3 | 408 | 40125997 | ||
BX094519.1 | - | 2, 3 | 272 | |||
D82070.1 | - | 2, 3 | 454 | |||
DB043858.1 | - | 3, 4 | 363 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623565.3 | - | 454 | Chr. - - [-] on Build |
