siRNA Details
| Gene Symbol: LINC01587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126517.1 | 1 | 274 | |||
| NR_126518.1 | 1 | 274 | ||||
| NR_126519.1 | 2 | 183 | ||||
| GenBank mRNA | BC117444.1 | - | 1 | 228 | 40125995 | |
| BC117446.1 | - | 1 | 228 | 40125997 | ||
| BX094519.1 | - | 1 | 92 | |||
| D82070.1 | - | 1 | 274 | |||
| DB043858.1 | - | 2 | 183 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000623565.3 | - | 274 | Chr. - - [-] on Build | ||

LINC01587
Genomic Map