siRNA Details
| Gene Symbol: PINK1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046507.1 | 1 | 2756 | |||
| GenBank mRNA | AK090466.1 | - | 1 | 2755 | ||
| AK172749.1 | - | 1 | 1842 | |||
| AY358957.1 | - | 1 | 135 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451424.1 | - | 1 | 2756 | Chr. 1 - 20642657 - 20652193 [-] on Build GRCh38 | |

PINK1-AS
Genomic Map