siRNA Details
| Gene Symbol: KIAA0087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_022006.1 | 1 | 175 | |||
| GenBank mRNA | D42038.1 | - | 1 | 138 | ||
| DB147006.1 | - | 1 | 175 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB171442.1 | - | 1 | 175 | Chr. 7 - 26536865 - 26538825 [-] on Build GRCh38 | |

KIAA0087
Genomic Map