siRNA Details
Gene Symbol: LINC00029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028295.1 | 1 | 733 | |||
GenBank mRNA | AL117533.1 | - | 1 | 733 | ||
BC069708.1 | - | 1 | 101 | |||
BC134343.1 | - | 1 | 58 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028295.1 | 1 | 733 | |||
GenBank mRNA | AL117533.1 | - | 1 | 733 | ||
BC069708.1 | - | 1 | 101 | |||
BC134343.1 | - | 1 | 58 |