siRNA Details
| Gene Symbol: LINC01260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034104.1 | 2, 3 | 300 | |||
| GenBank mRNA | BC000849.1 | - | 1, 2 | 165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01260| Gene Symbol: LINC01260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034104.1 | 2, 3 | 300 | |||
| GenBank mRNA | BC000849.1 | - | 1, 2 | 165 | ||