siRNA Details
| Gene Symbol: LINC00525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038407.1 | 1 | 12 | |||
| GenBank mRNA | BC007354.2 | - | 1 | 12 | 3616854 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412730.1 | - | 1 | 12 | Chr. 7 - 47761476 - 47766772 [+] on Build GRCh38 | |

LINC00525
Genomic Map