siRNA Details
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028509.1 | 2 | 1696 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000504876.2 | - | 2 | 1421 | Chr. 5 - 60488207 - 60526418 [+] on Build GRCh38 |
