siRNA Details
Gene Symbol: LOC150596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057517.1 | - | 2 | 1018 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000295052.3 | - | 1018 | Chr. 2 - 149858208 - 149858603 [-] on Build GRCh38 |
