siRNA Details
| Gene Symbol: C2orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182626.3 | NP_872432.1 | 4 | 693 | ||
| GenBank mRNA | AK057831.1 | - | 4 | 693 | ||
| BC093999.1 | - | 3 | 435 | 7939844 | ||
| BC094001.1 | - | 3 | 434 | 7939846 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000619640.3 | - | 404 | Chr. - - [-] on Build | ||

C2orf48
Genomic Map