siRNA Details
Gene Symbol: MIR202HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108078.1 | 3 | 750 | |||
NR_108079.1 | 2 | 602 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553459.1 | - | 2 | 608 | Chr. 10 - 133246478 - 133247891 [-] on Build GRCh38 |
