siRNA Details
Gene Symbol: SERHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027786.1 | 1 | 22 | |||
GenBank mRNA | AK094872.1 | - | 1 | 22 | ||
AL365513.1 | - | 11 | ||||
DA273263.1 | - | 1 | 22 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455578.5 | - | 1 | 22 | Chr. 22 - 42500579 - 42512559 [+] on Build GRCh38 |
