siRNA Details
| Gene Symbol: FLJ20712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012879.1 | XP_016868368.1 | 1 | 1093 | ||
| GenBank mRNA | AK000719.1 | - | 2 | 141 | ||
| AK225487.1 | - | 2 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ20712| Gene Symbol: FLJ20712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012879.1 | XP_016868368.1 | 1 | 1093 | ||
| GenBank mRNA | AK000719.1 | - | 2 | 141 | ||
| AK225487.1 | - | 2 | 141 | |||