siRNA Details
| Gene Symbol: LOC105370792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135681.1 | 3 | 722 | |||
| GenBank mRNA | AK057625.1 | - | 2 | 2854 | ||
| BC022404.1 | - | 1 | 905 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000309874.2 | - | 2 | 2854 | Chr. 15 - 41892793 - 41895935 [+] on Build GRCh38 | |

LOC105370792
Genomic Map