siRNA Details
| Gene Symbol: SNHG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003239.1 | 5 | 1070 | |||
| GenBank mRNA | AK055434.1 | - | 3 | 2636 | ||
| BC040237.1 | - | 3 | 2518 | |||
| Gene Symbol: SNORA60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091209.1 | - | 1 | 3069 | ||

SNHG11
Genomic Map