siRNA Details
| Gene Symbol: MIR1-1HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302812.1 | NP_001289741.1 | 5 | 565 | ||
| NM_178463.3 | NP_848558.1 | 4 | 946 | |||
| GenBank mRNA | AK056985.1 | - | 4 | 962 | ||
| BC110595.2 | - | 4 | 621 | 40022808 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000624914.3 | - | 946 | Chr. - - [-] on Build | ||

MIR1-1HG
Genomic Map