siRNA Details
| Gene Symbol: MIR1-1HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178463.3 | NP_848558.1 | 1 | 251 | ||
| GenBank mRNA | AK056985.1 | - | 1 | 251 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000624914.3 | - | 251 | Chr. - - [-] on Build | ||

MIR1-1HG
Genomic Map