siRNA Details
| Gene Symbol: LINC01106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR749351.1 | - | 1, 2 | 128 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436665.2 | - | 1, 2 | 128 | Chr. 2 - 110375138 - 110384442 [-] on Build GRCh38 | |

LINC01106
Genomic Map