siRNA Details
Gene Symbol: FLJ34503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027060.1 | 4 | 1322 | |||
GenBank mRNA | AK091822.1 | - | 4 | 1571 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000314481.3 | - | 1571 | Chr. 6 - 113904440 - 113919177 [+] on Build GRCh38 |
