siRNA Details
| Gene Symbol: LINC00302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF005082.1 | - | 1, 2 | 63 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444515.1 | - | 1, 2 | 63 | Chr. 1 - 152655477 - 152656804 [+] on Build GRCh38 | |

LINC00302
Genomic Map