siRNA Details
| Gene Symbol: LINC00982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015440.1 | 3 | 3247 | |||
| GenBank mRNA | AK124865.1 | - | 3 | 1763 | ||
| AL713743.1 | - | 1 | 1244 | |||
| AL832943.1 | - | 3 | 2901 | |||
| AL833006.1 | - | 1 | 744 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00982| Gene Symbol: LINC00982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015440.1 | 3 | 3247 | |||
| GenBank mRNA | AK124865.1 | - | 3 | 1763 | ||
| AL713743.1 | - | 1 | 1244 | |||
| AL832943.1 | - | 3 | 2901 | |||
| AL833006.1 | - | 1 | 744 | |||