siRNA Details
| Gene Symbol: C8orf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173687.3 | NP_775958.1 | 6 | 1295 | ||
| NR_126398.1 | 8 | 1477 | ||||
| NR_126399.1 | 6 | 1286 | ||||
| GenBank mRNA | AK094450.1 | - | 6 | 1243 | ||
| BC120993.2 | - | 6 | 1221 | |||

ENSG00000177335
Genomic Map