siRNA Details
| Gene Symbol: FLJ40194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034161.1 | 2 | 466 | |||
| GenBank mRNA | AK097513.1 | - | 2 | 470 | ||
| BI463223.1 | - | 2 | 471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ40194| Gene Symbol: FLJ40194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034161.1 | 2 | 466 | |||
| GenBank mRNA | AK097513.1 | - | 2 | 470 | ||
| BI463223.1 | - | 2 | 471 | |||