siRNA Details
| Gene Symbol: LOC100049716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122124.1 | 5 | 1129 | |||
| GenBank mRNA | AK056115.1 | - | 5 | 1129 | ||
| BC033133.1 | - | 2 | 1448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100049716| Gene Symbol: LOC100049716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122124.1 | 5 | 1129 | |||
| GenBank mRNA | AK056115.1 | - | 5 | 1129 | ||
| BC033133.1 | - | 2 | 1448 | |||