siRNA Details
| Gene Symbol: LOC648987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034086.1 | 2 | 1905 | |||
| NR_104649.1 | 3 | 2021 | ||||
| NR_104650.1 | 2 | 275 | ||||
| NR_104651.1 | 3 | 380 | ||||
| GenBank mRNA | BC013653.1 | - | 2 | 1819 | ||
| BC039088.1 | - | 3 | 2021 | |||
| BC046188.1 | - | 2 | 1818 | |||
| BF130969.1 | - | 356 | ||||
| HY041253.1 | - | 2 | 276 | |||

LOC648987
Genomic Map