siRNA Details
| Gene Symbol: LOC90768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027107.2 | 4 | 1412 | |||
| GenBank mRNA | AK024300.1 | - | 4 | 1363 | ||
| BC033172.1 | - | 1 | 864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC90768| Gene Symbol: LOC90768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027107.2 | 4 | 1412 | |||
| GenBank mRNA | AK024300.1 | - | 4 | 1363 | ||
| BC033172.1 | - | 1 | 864 | |||