siRNA Details
| Gene Symbol: LOC90768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056196.1 | - | 5 | 1703 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000507869.5 | - | 5 | 1703 | Chr. 4 - 181975156 - 182085136 [-] on Build GRCh38 | |

LOC90768
Genomic Map