siRNA Details
| Gene Symbol: C9orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173520.2 | NP_775791.1 | 3 | 1023 | ||
| GenBank mRNA | BC034752.1 | - | 3 | 1023 | 5193354 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000623103.1 | - | 1023 | Chr. - - [-] on Build | ||

C9orf62
Genomic Map