siRNA Details
Gene Symbol: LOC284241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136643.1 | 5 | 1094 | |||
GenBank mRNA | AK098581.1 | - | 5 | 1093 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136643.1 | 5 | 1094 | |||
GenBank mRNA | AK098581.1 | - | 5 | 1093 |