siRNA Details
Gene Symbol: LINC00324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026951.1 | 2 | 715 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000315707.3 | - | 2 | 715 | Chr. 17 - 8220667 - 8224043 [-] on Build GRCh38 |
