siRNA Details
| Gene Symbol: LINC00670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034144.1 | 2 | 361 | |||
| NR_034145.1 | 3 | 523 | ||||
| GenBank mRNA | AK092009.1 | - | 3 | 522 | ||
| BC122562.1 | - | 2 | 348 | |||
| BC122870.1 | - | 2 | 348 | |||
| DB255288.1 | - | 3 | 524 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000577679.1 | - | 2 | 348 | Chr. 17 - 12549981 - 12635782 [+] on Build GRCh38 | |

LINC00670
Genomic Map