siRNA Details
Gene Symbol: LINC00670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034144.1 | 3 | 1049 | |||
NR_034145.1 | 4 | 1211 | ||||
GenBank mRNA | AK092009.1 | - | 4 | 1210 | ||
BC122562.1 | - | 3 | 1036 | |||
BC122870.1 | - | 3 | 1036 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000577679.1 | - | 3 | 1036 | Chr. 17 - 12549981 - 12635782 [+] on Build GRCh38 |
