siRNA Details
| Gene Symbol: LINC00305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027245.1 | 4 | 547 | |||
| GenBank mRNA | AY336744.1 | - | 3 | 310 | ||
| BC029565.1 | - | 4 | 551 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00305| Gene Symbol: LINC00305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027245.1 | 4 | 547 | |||
| GenBank mRNA | AY336744.1 | - | 3 | 310 | ||
| BC029565.1 | - | 4 | 551 | |||