siRNA Details
| Gene Symbol: C10orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173541.2 | NP_775812.1 | 3 | 673 | ||
| GenBank mRNA | BC030794.1 | - | 3 | 694 | 3898116 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000321248.4 | - | 3 | 694 | Chr. 10 - 132445189 - 132449386 [+] on Build GRCh38 | |

C10orf91
Genomic Map