siRNA Details
| Gene Symbol: LOC728175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040108.1 | 3 | 1570 | |||
| GenBank mRNA | AK097110.1 | - | 3 | 1570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728175| Gene Symbol: LOC728175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040108.1 | 3 | 1570 | |||
| GenBank mRNA | AK097110.1 | - | 3 | 1570 | ||