siRNA Details
| Gene Symbol: C17orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302809.1 | NP_001289738.1 | 3 | 899 | ||
| NM_152460.2 | NP_689673.2 | 3 | 735 | |||
| GenBank mRNA | AK056444.1 | - | 3 | 735 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17orf77| Gene Symbol: C17orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302809.1 | NP_001289738.1 | 3 | 899 | ||
| NM_152460.2 | NP_689673.2 | 3 | 735 | |||
| GenBank mRNA | AK056444.1 | - | 3 | 735 | ||