siRNA Details
Gene Symbol: LINC01006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103858.1 | 2 | 189 | |||
GenBank mRNA | BM793926.1 | - | 2 | 189 | ||
BU932175.1 | - | 4 | 370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425712.1 | - | 2 | 189 | Chr. 7 - 156472196 - 156603195 [-] on Build GRCh38 |
