siRNA Details
Gene Symbol: COL18A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028082.1 | 1 | 198 | |||
GenBank mRNA | BC111539.2 | - | 1 | 198 | 40026216 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: COL18A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028082.1 | 1 | 198 | |||
GenBank mRNA | BC111539.2 | - | 1 | 198 | 40026216 |