siRNA Details
| Gene Symbol: C2orf27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214461.2 | NP_999626.1 | 2 | 383 | ||
| GenBank mRNA | BC043584.1 | - | 2 | 383 | 5229449 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000623129.1 | - | 383 | Chr. - - [-] on Build | ||

C2orf27B
Genomic Map