siRNA Details
| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 5 | 664 | |||
| NR_028410.1 | 4 | 633 | ||||
| NR_028411.1 | 4 | 597 | ||||
| GenBank mRNA | BC144506.1 | - | 4 | 621 | ||
| BC144508.1 | - | 5 | 652 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENSG00000187012
LINC00207| Gene Symbol: LINC00207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028409.1 | 5 | 664 | |||
| NR_028410.1 | 4 | 633 | ||||
| NR_028411.1 | 4 | 597 | ||||
| GenBank mRNA | BC144506.1 | - | 4 | 621 | ||
| BC144508.1 | - | 5 | 652 | |||