siRNA Details
| Gene Symbol: LINC00615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038868.1 | 3 | 267 | |||
| GenBank mRNA | AK058107.1 | - | 3 | 267 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000546725.1 | - | 3 | 267 | Chr. 12 - 90918023 - 90948669 [+] on Build GRCh38 | |

LINC00615
Genomic Map