siRNA Details
| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 7 | 649 | |||
| NR_110307.1 | 6 | 496 | ||||
| NR_110308.1 | 7 | 668 | ||||
| GenBank mRNA | BC033546.1 | - | 7 | 639 | ||
| BC070365.1 | - | 7 | 629 | |||
| BI829888.1 | - | 6 | 460 | |||

LINC00238
Genomic Map